missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Super Biohazard Disposal Bags

Protects against dangerous tears and leaks. Super Biohazard Disposal Bags feature a sterilization indicator patch that darkens when exposed to steam sterilization for 20 minutes.

Brand:  Bel-Art™ F13165-1923

321.21 EUR valid until 2025-05-09
Use promo code "24143" to get your promotional price.


 View more versions of this product

Product Code. 17672085

  • 422.00 € / Pack of 200

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Protect against dangerous tears and leaks
  • Orange-red bags have standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish
  • Extra-heavy-duty bags are 2 mil-thick blended polymer
  • Autoclavable to 121°C (250°F)
  • Sterilization indicator patch darkens when exposed to steam sterilization
Specifications

Specifications

19 L
5.0 gal.
23 x 19 in. (58 x 48 cm)
58 cm
48 cm
Standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish
19 in.
2 mil
Biohazard Disposal Bag
Zip Tie
Red
Single-use
Non-sterile
Autoclavable
Extra-strength, high-impact, high-temperature bags reduce autoclave time
23 in.
Polymer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Super Biohazard Disposal Bags > 19 x 23 in. (48 x 58 cm)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.