missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Super Biohazard Disposal Bags

Protects against dangerous tears and leaks. Super Biohazard Disposal Bags feature a sterilization indicator patch that darkens when exposed to steam sterilization for 20 minutes.

291.00 € - 1020.00 €

Specifications

Color Red
Autoclavable Autoclavable
Includes Standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish
For Use With (Application) Extra-strength, high-impact, high-temperature bags reduce autoclave time
Thickness 2 mil
View More Specs

Products 5
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
17662085
View Documents
Bel-Art™
F13165-1419
19 x 14 in. (48 x 36 cm)
291.00 €
Pack of 200
Please to purchase this item. Need a web account? Register with us today!
10287083
View Documents
Bel-Art™
F13165-2535
35 x 25 in. (35 x 64 cm)
745.06 €
Pack of 200
Please to purchase this item. Need a web account? Register with us today!
17672085
View Documents
Bel-Art™
F13165-1923
23 x 19 in. (58 x 48 cm)
422.00 €
Pack of 200
Please to purchase this item. Need a web account? Register with us today!
17692085
View Documents
Bel-Art™
F13165-3748
37 x 48 in. (94 x 122 cm)
733.00 €
Pack of 100
Estimated Shipment: 02-10-2025
to see stock.
Add to basket
 
17682085
View Documents
Bel-Art™
F13165-3138
31 x 38 in. (79 x 96 cm)
1020.00 €
Pack of 200
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Protect against dangerous tears and leaks
  • Orange-red bags have standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish
  • Extra-heavy-duty bags are 2 mil-thick blended polymer
  • Autoclavable to 121°C (250°F)
  • Sterilization indicator patch darkens when exposed to steam sterilization
Specifications

Specifications

Red
Standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish
2 mil
Biohazard Disposal Bag
Autoclavable
Extra-strength, high-impact, high-temperature bags reduce autoclave time
Polymer
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Super Biohazard Disposal Bags

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.