Learn More
Description
The most important of the provitamins A, β-carotene can be classified as an antioxidant due to its inhibition of radical initiated peroxidation in vitro.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
The most important of the provitamins A, β-carotene can be classified as an antioxidant due to its inhibition of radical initiated peroxidation in vitro.
Solubility
Soluble in hexane, dimethyl sulfoxide, benzene, chloroform, cyclohexane. Insoluble in water.
Notes
Air & Light Sensitive. Store in a cool place.

Specifications
Specifications
Melting Point | 178°C to 179°C |
Density | 1.000 g/mL |
Flash Point | 103°C (217°F) |
Quantity | 5 g |
Beilstein | 1917416 |
Sensitivity | Air and light sensitive |
Solubility Information | Soluble in hexane,dimethyl sulfoxide,benzene,chloroform,cyclohexane. Insoluble in water. |
Formula Weight | 536.87 |
Percent Purity | 99% |
Physical Form | Solid |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T30K026 | Certificate of Analysis | 13/08/2024 | H60106.03,H60106.06 |
S28J044 | Certificate of Analysis | 24/07/2023 | H60106.03,H60106.06 |
W30I002 | Certificate of Analysis | 13/10/2022 | H60106.03,H60106.06 |
R25E052 | Certificate of Analysis | 23/11/2021 | H60106.03,H60106.06 |
P10B033 | Certificate of Analysis | 23/11/2021 | H60106.03,H60106.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.