missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Biotin Hydrazide

MedChemExpress Biotin Hydrazide is a carbonyl-reactive biotinylation reagent, which is a carbonyl probe.

Brand:  MedChemExpress HY-100215

Encompass_Preferred

Product Code. 16484256

  • 83.70 € / 100mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

100 mg
Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
66640-86-6
DMSO : 6.6 mg/mL (25.55 mM; Need ultrasonic and warming) ∣ H2O : 2 mg/mL (7.74 mM; Need ultrasonic)
Biochemical Assay Reagents
Room temperature
258.34 g/mol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
MedChemExpress Biotin Hydrazide

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.