missing translation for 'onlineSavingsMsg'
Learn More
Bis(tri-n-butyltin) Oxide, Stabilized 96%, Thermo Scientific™ Product Code.: 10539810

Bis(tri-n-butyltin) Oxide, Stabilized 96%, Thermo Scientific™

Product Code. 10539810
100 mL, Glass bottle
Click to view available options
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10539810

Brand: Thermo Scientific Acros 106511000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-35-9
Molecular Formula C24H58OSn2
Molecular Weight (g/mol) 600.15
MDL Number MFCD00009418
InChI Key NZQFYJHTWVEUPP-UHFFFAOYSA-N
Synonym tributyltin oxide, tbto, bis tributyltin oxide, hexabutyldistannoxane, distannoxane, hexabutyl, lastanox q, bis tri-n-butyltin oxide, butinox, biometShow MoreShow Less
PubChem CID 16682746
ChEBI CHEBI:81543
IUPAC Name tributyltin oxide
SMILES O.CCCC[SnH](CCCC)CCCC.CCCC[SnH](CCCC)CCCC

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula [CH3(CH2)3]3SnOSn[(CH2)3CH3]3
Refractive Index 1.4854 to 1.4874 (20°C, 589nm)
Quantity 100 mL
Fieser 06,56; 07,26; 08,43; 09,53; 11,62; 13,41; 16,227
Stabilizer 0.1% lactic acid
Formula Weight 596.11
Percent Purity ≥95% (ex Sn)
Physical Form Liquid
Chemical Name or Material Bis(tri-n-butyltin) oxide, 96%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bis(tri-n-butyltin) Oxide, Stabilized 96%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.