Learn More
BRAND™ BLAUBRAND™ Bang Pattern Micro Burette
With interchangeable straight needle-valve stopcock with PTFE spindle (PTFE key in the intermediate stopcock)
Brand:  BRAND™ 24267
 View more versions of this product
Product Code. 15291919
Description
Description
  • Class AS
  • Boro 3.3 Glass
  • To DIN EN ISO 385
  • Calibrated to deliver (TD, Ex)
  • Schellbach stripe
  • All BLAUBRAND™ burettes are delivered with batch number and enclosed one batch certificate per pack
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    5 mL
    0.01 mL
    0.01 mL
    Borosilicate Glass (3.3)
    2
    Class AS
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    BRAND™ BLAUBRAND™ Bang Pattern Micro Burette > Capacity: 5 mL