Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Compact Burettes, with Certificate
Fast to dismantle and easy to clean. BRAND™ Blaubrand™ Borosilicate Glass Compact Burettes are also simple to repair because all individual components replaceable. Delivered with individual conformity certificate.
Brand:  BRAND™ 913916
99.15 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15884958
  • 126.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Includes removable PTFE stopcock
  • Fast to dismantle and easy to clean.
  • Simple repair - all individual components replaceable.
  • Borosilicate glass construction (Boro 3.3 to DIN EN ISO 385.)
  • Calibrated to deliver (TD, Ex).
  • Schellbach stripe.
  • Delivered with individual conformity certificate.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
250 mL
Borosilicate Glass
DIN EN ISO 385
800 mm
±0.03 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BRAND™ BLAUBRAND™ Borosilicate Glass Compact Burettes, with Certificate > Capacity: 25 mL