Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Micro Burettes with PTFE Spindle
Constructed from borosilicate glass. BRAND™ Blaubrand™ Borosilicate Glass Micro Burettes with PTFE Spindle are calibrated to deliver contents that correspond to a volume as specified.
317.22 € - 346.80 €
Specifications
Material | Borosilicate Glass |
---|---|
Includes | Schellbach Stripes |
Stopcock | Lateral |
Stopcock Material | Glass |
Class | Class AS |
Description
- Borosilicate glass construction (Boro 3.3. To DIN EN ISO 385)
- Calibrated to deliver (TD, Ex)
- Bang pattern
- Schellbach stripe
- Includes interchangeable needle-valve stopcock with PTFE spindle
- Stand included
- All BLAUBRAND™ burettes are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer
- On demand also available with individual certificate, USP Individual certificate or DAkkS calibration certificate
Specifications
Borosilicate Glass | |
Lateral | |
Class AS |
Schellbach Stripes | |
Glass | |
DIN EN ISO 385 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BRAND™ BLAUBRAND™ Borosilicate Glass Micro Burettes with PTFE Spindle