Learn More
BRAND™ Blaubrand™ Class A Borosilicate Glass Volumetric Flask with Polypropylene Stopper
Constructed of Borosilicate Glass and complies with DIN EN ISO 1042 standards. BRAND™ Blaubrand™ Class A Borosilicate Glass Volumetric Flask with Polypropylene Stopper is available with narrow neck or wide neck in a variety of sizes. DE-M marking on the neck provides quick and accurate volume check. Polypropylene stopper included.
Brand: BRAND™ 37241
36.40 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Class A Volumetric Flask
- Constructed of Borosilicate Glass
- Complies with DIN EN ISO 1042 standards
- DAkkS calibration certificate available upon request
- Calibrated to contain (TC, In)
- Narrow neck and wide neck options available in a variety of sizes
- DE-M marking provides quick and accurate volume check
- Polypropylene stopper included

Specifications
Ground Joint NS 7/16 | |
Polypropylene stopper | |
10 mL |
Product Suggestions
Customers who viewed this item also viewed.
Videos
-
Brand BLAUBRAND
Your input is important to us. Please complete this form to provide feedback related to the content on this product.