missing translation for 'onlineSavingsMsg'
Learn More
BOC-L-Methionine, 98%, Thermo Scientific Chemicals Product Code.: 10723061

BOC-L-Methionine, 98%, Thermo Scientific Chemicals

Product Code. 10723061
5 g, Glass Bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10723061

Brand: Thermo Scientific Acros 275630050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2488-15-5
Molecular Formula C10H19NO4S
Molecular Weight (g/mol) 249.33
MDL Number MFCD00065586
InChI Key IMUSLIHRIYOHEV-ZETCQYMHSA-N
Synonym boc-l-methionine, boc-met-oh, n-boc-l-methionine, n-tert-butoxycarbonyl-l-methionine, tert-butoxycarbonyl-l-methionine, s-2-tert-butoxycarbonyl amino-4-methylthio butanoic acid, n-alpha-t-butyloxycarbonyl-l-methionine, methionine, n-1,1-dimethylethoxy carbonyl, l-methionine, n-1,1-dimethylethoxy carbonylShow MoreShow Less
PubChem CID 89857
SMILES CSCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 47.0°C to 53.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula CH3SCH2CH2CH(COOH)NHCOOC(CH3)3
Specific Rotation − 23.00
Water 0.5% max. (K.F.)
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Quantity 5 g
Formula Weight 249.33
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material BOC-L-Methionine, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number345256 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number275630250,275630050
Lot Number345256 Certificate TypeCertificate of Analysis Date11/07/2024 Catalog Number275630250,275630050
Lot Number344202 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number275630250,275630050
Lot Number344036 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number275630250,275630050
Lot Number344396 Certificate TypeCertificate of Analysis Date02/09/2020 Catalog Number275630250,275630050
Product Identifier
  • BOC-L-Methionine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BOC-L-Methionine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.