missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Boekel Glass/Stainless Steel Desiccator
Keep samples secure in this desiccator with double-action lock. Cole-Parmer™ Boekel Glass/Stainless Steel Desiccators feature airtight, leakproof door with double-action lock, one-piece neoprene gasket and full-length hinge.
1620.00 € - 4380.00 €
Specifications
Material | Stainless Steel, Glass |
---|---|
Product Type | Desiccator |
No. of Shelves | 2 |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
11796118
|
Cole-Parmer™
24000-10 |
125 L |
4380.00 €
Each |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
11706128
|
Cole-Parmer™
08901-60 |
22 L | N/A | |||||||
Description
- Equalize pressure with manual pressure release valve
- Manual pressure relief valve lets you equalize internal and external pressure when opening door.
- Stainless steel desiccant tray holds one pound of desiccant
- High-polish 302 stainless steel cabinets with 3/16 in. thick glass door and sides for observing contents
- Door has double-action lock, one-piece rubber gasket and full-length hinge
- Manual pressure vent valve lets you release pressure
- Choose from flexboard or SS shelves
- SS desiccant tray holds one pound of desiccant
Specifications
Stainless Steel, Glass | |
Desiccator | |
2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ Boekel Glass/Stainless Steel Desiccator