missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Boldenone
Brand: Thermo Scientific™ 465751000
125.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
This item is not returnable.
View return policy
Description

Specifications
Boldenone | |
100 mg | |
C19 H26 O2 | |
Powder or Crystals |
846-48-0 | |
168°C to 170°C | |
286.41 | |
White to Light-yellow to Light-beige |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0419244 | Certificate of Analysis | 08/12/2022 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Boldenone > 100 mg
Spot an opportunity for improvement?Share a Content Correction