Learn More
Scientific Glass Labs™ Borosilicate Glass Bell Jar with Socket Top
Borosilicate glass construction. Includes socket top without stopper.
Brand:  Scientific Glass Labs™ 2874/300X200
 View more versions of this product
Product Code. 12706899
Description
Description
  • Borosilicate glass construction
  • Clear
  • Socket top for stopper
  • Ground Flange
Specifications
Specifications
Socket Top
Clear
300 mm
Bell Jar
0.2 kg
185 mm
Borosilicate Glass
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Scientific Glass Labs™ Borosilicate Glass Bell Jar with Socket Top > Outer Diameter: 185mm; with 29/32 Socket Top