Learn More
Cytiva Whatman™ UNIPLATE™ Microplates
Manufactured from polypropylene for superior chemical and thermal stability. Whatman™ UNIPLATE™ Microplates are suitable for simple filtrate collection and homogeneous assay techniques used in high-throughput screening (HTS).
Brand: Cytiva 7701-5110
319.92 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Manufactured from polypropylene for superior chemical and thermal stability. Whatman™ UNIPLATE™ Microplates are suitable for simple filtrate collection and homogeneous assay techniques used in high-throughput screening (HTS).
- Withstand low temperatures (-80°C) for long-term storage
- Conform to ANSI™/SBS standards for compatibility with most microplate readers and automated plate-handling devices

Specifications
Standard | |
UNIPLATE microplates | |
Natural | |
10 mL | |
Sterile | |
Ideal for homogeneous assay techniques utilized in high-throughput screening |
With Lid | |
4 x 6 | |
Polypropylene | |
24 | |
Round | |
25/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.