missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug

For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug are manufactured from durable DURAN™ borosilicate glass.

Brand:  Lenz Laborglasinstrumente™ 04004137

 View more versions of this product

Product Code. 11925966

  • 50.30 € / Pack of 2
Estimated Shipment: 23-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Without scale
  • With NS socket and PE stopper
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications

Specifications

100 mL
No.
2
Borosilicate Glass
2.5mm bore
Polyethylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug > Capacity: 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.