Learn More
Brady™ BBP™ Hi-Performance Polyester Tape
BBP™ Hi-Performance Polyester Tape (B-569) has low halide content and is designed for marking stainless steel pipes particularly in harsh environments.
148.00 € - 477.00 €
Specifications
Adhesive Type | Acrylic, Permanent, Pressure-sensitive |
---|---|
Finish | Gloss |
Format | Tape |
Material | Polyester |
Sterility | Non-sterile |
Product Code | Brand | Color | Width (English) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Width (English) | Price | Quantity & Availability | ||||||
11776385
|
Brady™
117930 |
Black | 2.25 in. |
327.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11796385
|
Brady™
117932 |
Black | 0.5 in. |
152.00 €
Each |
In Stock
Log in to see stock. |
||||||
11766385
|
Brady™
117929 |
Black | 4 in. |
477.00 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
11786385
|
Brady™
117931 |
Black | 1.12 in. |
228.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11706395
|
Brady™
117933 |
Blue | 4 in. |
477.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11716395
|
Brady™
117934 |
Blue | 2.25 in. |
327.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11726395
|
Brady™
117935 |
Blue | 1.12 in. |
228.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11736395
|
Brady™
117936 |
Blue | 0.5 in. |
152.00 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
11706405
|
Brady™
117943 |
Clear | 1.12 in. |
228.00 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
11786395
|
Brady™
117941 |
Clear | 4 in. |
477.00 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
11716405
|
Brady™
117944 |
Clear | 0.5 in. | N/A | N/A | N/A | |||||
11796395
|
Brady™
117942 |
Clear | 2.25 in. |
327.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11726405
|
Brady™
117945 |
Green | 4 in. |
477.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11736405
|
Brady™
117946 |
Green | 2.25 in. |
327.00 €
Each |
In Stock
Log in to see stock. |
||||||
11746405
|
Brady™
117947 |
Green | 1.12 in. |
228.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11756405
|
Brady™
117948 |
Green | 0.5 in. |
152.00 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
11706425
|
Brady™
117963 |
Orange | 1.12 in. |
228.00 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
11786415
|
Brady™
117961 |
Orange | 4 in. |
477.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11716425
|
Brady™
117964 |
Orange | 0.5 in. |
152.00 €
Each |
In Stock
Log in to see stock. |
||||||
11796415
|
Brady™
117962 |
Orange | 2.25 in. |
327.00 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
11706435
|
Brady™
117973 |
Red | 4 in. |
477.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11716435
|
Brady™
117974 |
Red | 2.25 in. |
327.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11726435
|
Brady™
117975 |
Red | 1.12 in. |
228.00 €
Each |
In Stock
Log in to see stock. |
||||||
11736435
|
Brady™
117976 |
Red | 0.5 in. |
152.00 €
Each |
In Stock
Log in to see stock. |
||||||
11746445
|
Brady™
117987 |
White | 2.25 in. |
327.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11726445
|
Brady™
117985 |
White | 4 in. |
477.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11736445
|
Brady™
117986 |
White | 3 in. |
428.00 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
11766445
|
Brady™
117989 |
White | 0.5 in. |
152.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11756445
|
Brady™
117988 |
White | 1.12 in. |
228.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11776445
|
Brady™
117990 |
Yellow | 4 in. |
442.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11786445
|
Brady™
117991 |
Yellow | 2.25 in. |
327.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11796445
|
Brady™
117992 |
Yellow | 1.125 in. |
228.00 €
Each |
In Stock
Log in to see stock. |
||||||
11706455
|
Brady™
117993 |
Yellow | 0.5 in. |
152.00 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
Description
- Durable, ultra-smooth polyester labels with permanent pressure sensitive acrylic adhesive
- Outdoor grade polyester tape can be thermal transfer printed
- Withstands mild chemicals, oil and water
- Enables printers with XY cutters to cut custom shapes or multiple labels out of a single continuous roll
Specifications
Acrylic, Permanent, Pressure-sensitive | |
Tape | |
Non-sterile | |
BBP30, BBP31, BBP33, BBP35, and BBP37 printers | |
3 to 5 Years | |
-40 to 212°F | |
45 lb./in. | |
100 ft. |
Gloss | |
Polyester | |
Designed for use on Stainless Steel pipes in harsh environments such as the pulp and Paper industry | |
80 Gardner Units | |
1 Roll | |
-40°C to +100°C | |
0.003 in. | |
30.48 m |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Brady™ BBP™ Hi-Performance Polyester Tape