Learn More
Brady™ Label Maker Vinyl Cartridges (B-427) for BMP51, BMP53, BMP41 Printers
Designed to adhere to highly curved surfaces, wires and cables
32.25 € - 85.15 €
Specifications
Material Self-laminating Vinyl (B-427)
Product Type Label Maker Cartridge
For Use With (Equipment) BMP™51,BMP™53,BMP™41 Printers
View More Specs
Products 15
Product Code Brand Color Size (Metric) Size (English) Price Quantity & Availability  
Product Code Brand Color Size (Metric) Size (English) Price Quantity & Availability  
15103339
View Documents
Brady™
131574
Black, White, Clear 12.7 x 38.1 mm 0.5 x 1.5 in. N/A N/A N/A
15153339
View Documents
Brady™
131573
Black, Yellow 25.4 x 38.1 mm 1 x 1.5 in. N/A N/A N/A
15133339
View Documents
Brady™
131570
Black, White, Clear 25.4 x 38.1 mm 1 x 1.5 in. N/A N/A N/A
15113339
View Documents
Brady™
131572
Black, White, Clear 19.050 x 38.1 mm 0.75 x 1.5 in. N/A N/A N/A
15143339
View Documents
Brady™
131601
Black, Yellow 25.4 x 101.6 mm 1 x 4 in. N/A N/A N/A
15163329
View Documents
Brady™
131579
Black, White, Clear 25.4 x 63.5 mm 1 x 2.5 in. N/A N/A N/A
15133329
View Documents
Brady™
131584
Black, White, Clear 19.050 x 25.4 mm 0.75 x 1 in. N/A N/A N/A
15123339
View Documents
Brady™
131585
Black, Yellow 25.4 x 63.5 mm 1 x 2.5 in. N/A N/A N/A
15173329
View Documents
Brady™
131587
Black, White, Clear 25.4 x 101.6 mm 1 x 4 in. N/A N/A N/A
15153329
View Documents
Brady™
131580
Black, White, Clear 25.4 x 25.4 mm 1 x 1 in. N/A N/A N/A
15143329
View Documents
Brady™
131589
Black, White, Clear 12.7 x 25.4 mm 0.5 x 1 in. N/A N/A N/A
15193329
View Documents
Brady™
131571
Black, White, Clear 25.4 x 31.750 mm 1 x 1.25 in. N/A N/A N/A
15123329
View Documents
Brady™
131581
Black, White, Clear 25.4 x 19.050 mm 1 x 0.75 in. N/A N/A N/A
11942782
View Documents
Brady™
143261
Black, White, Clear 12.7 x 31.75 mm 0.5 x 1.25 in. N/A N/A N/A
11972772
View Documents
Brady™
143252
Black, White, Clear 12.7 x 19.050 mm 0.5 x 0.75 in. N/A N/A N/A
Description
Description
  • Feature printable zone in white or yellow
  • Transparent, non printable area wraps around wires and cables for laminating printed texts
  • Offers resistance to abrasion, handling, solvents, water, oil and dirt
  • Available in various sizes
Specifications
Specifications
Self-laminating Vinyl (B-427)
Label Maker Cartridge
BMP™51,BMP™53,BMP™41 Printers
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Brady™ Label Maker Vinyl Cartridges (B-427) for BMP51, BMP53, BMP41 Printers