Learn More
BRAND™ 96-well Deep-well Plates
These deep-well plates are stackable for easy storage.
Brand: BRAND™ 701347
239.69 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
- DNA-, DNase- and RNase-free
- Designed for a wide range of applications, such as High-Throughput Screening (HTS), tests requiring a mother plate, cell and tissue cultures, serial dilutions, reagent transfer and sample storage down to -80°C
- Deep-well plates are manufactured according to ANSI/SLAS 1 and 4 in.
- Choose PP plates for autoclavability and chemical resistance.
- Choose PS plates for transparency and surface properties.
- Deep Well Plates are ideal for sample storage, HTS assays requiring a motherplate, cell and tissue culture, immunological assays, and other applications.

Specifications
No Barcode | |
Standard | |
8 x 12 | |
Clear | |
Deep-well Plate | |
Polypropylene | |
96 | |
Square, Round Bottom (U-well) | |
45 cm | |
14 cm |
ANSI/SLAS, Certified Life Science Quality, BIO-CERT™ PCR Quality | |
96-well Deep-well plate, PP, well shape square, U-bottom, Low Profile, 1 ml | |
Autoclavable | |
Single-use | |
PCR Machine | |
1 mL | |
Non-sterile | |
Low Profile | |
28 cm | |
PCR Ready |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
20230105 | Certificate of Analysis | 10/11/2023 | 701347 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.