Learn More
Cole Parmer BrandTech Pipette Controller
BrandTech Pipette Controller is color-coded for quick identification
Brand:  Cole Parmer 24550-49
Product Code. 15715072
  • 464.00 € / Each
Estimated Shipment: 30-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
BrandTech Pipette Controller is color-coded for quick identification
  • Measure and dispense with a single lever
  • Blow-out button clears the pipette
  • Adapter fits all pipettes from 0.1 to 100 mL
  • Membrane filter protects controller from fumes and liquids
  • Filter and adapter may be autoclaved
Specifications
Specifications
Natural
Non-sterile
100 mL
BrandTech
Pipette Controller
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole Parmer BrandTech Pipette Controller > Color: Natural