missing translation for 'onlineSavingsMsg'
Learn More

Branson Ultrasonics™ CPX Series Ultrasonic Cleaning Bath

Outfit your lab with a classic tabletop cleaner, offering high-tech design, solid construction and featuring reliable digital controls with power tracking capabilities.

Brand:  Branson Ultrasonics™ CPX-952-239R

 View more versions of this product

Product Code. 13423619

  • 1346.80 € / Each
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

LCD
90 min.
230/240 V
2.8 L
Cleaning
CPX2800-E
4.1 kg
Plastic
40 kHz
0.75 gal.
23 x 14 x 10 cm (9.5 x 5.5 x 4 in.)
Tank cover
9 lb.
2 Years
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Branson Ultrasonics™ CPX Series Ultrasonic Cleaning Bath > CPX2800-E; 230/240V; Capacity: 2.8L; 33.7L x 30.4W x 30.2cm H

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.