Learn More
Brady™ Breaker Blocker Component - Green Blocking Bar
Helps to prevent electrical risk.
Brand:  Brady™ 090893
Product Code. 15643517
  • 34.80 € / Pack of 5
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Green bar indicates locked ‘on’
Specifications
Specifications
Breaker Blocker Components
Green
5 Pk.
EZ 480/600 VOLT 7.6 in. GREEN BLOCK BAR PK5
Breaker Blocker Components - 190 mm Green Blocking Bar
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ Breaker Blocker Component - Green Blocking Bar > Green