missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
Color | Blue |
pH | 10 |
Quantity | 50 Capsules |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer Capsules pH 10, Reagecon™ >
Spot an opportunity for improvement?Share a Content Correction