Learn More
Buffer solution pH 5, Acetate buffer, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 258620010
46.11 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
pH buffer
Specifications
Buffer solution pH 5 | |
Colorless | |
4.98 to 5.02 (20°C) | |
1 L | |
MFCD03097943 | |
1 |
64-19-7, 7732-18-5, 1310-58-3 | |
1.0000g/mL | |
Liquid | |
0.7mL glacial acetic acid and 0.6g potassium hydroxide per liter | |
Plastic bottle | |
Solubility in water: soluble. |
Safety and Handling
- Buffer solution pH 5, Acetate buffer
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0464202 | Certificate of Analysis | 28/06/2024 | 258620010 |
A0457509 | Certificate of Analysis | 15/11/2023 | 258620010 |
A0450686 | Certificate of Analysis | 02/05/2023 | 258620010 |
A0451365 | Certificate of Analysis | 02/05/2023 | 258620010 |
A0401277 | Certificate of Analysis | 15/12/2022 | 258620010 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only