missing translation for 'onlineSavingsMsg'
Learn More
Butadiene sulfone, 98%, Thermo Scientific Chemicals Product Code.: 15169163

Butadiene sulfone, 98%, Thermo Scientific Chemicals

Product Code. 15169163
2.5 kg, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 15169163

Brand: Thermo Scientific Acros 107610025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-79-2
Molecular Formula C4H6O2S
Molecular Weight (g/mol) 118.16
MDL Number MFCD00005481
InChI Key MBDNRNMVTZADMQ-UHFFFAOYSA-N
Synonym 3-sulfolene, butadiene sulfone, sulfolene, thiophene, 2,5-dihydro-, 1,1-dioxide, beta-sulfolene, 2,5-dihydrothiophene sulfone, 2,5-dihydrothiophene dioxide, 2,5-dihydrothiophene-1,1-dioxide, sulfol-3-ene, ccris 569Show MoreShow Less
PubChem CID 6498
IUPAC Name 2,5-dihydrothiophene 1,1-dioxide
SMILES C1C=CCS1(=O)=O

Specifications

Melting Point 63.0°C to 66.0°C
Color Brown or Orange to Yellow
Density 1.3000g/mL
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Quantity 2.5 kg
Beilstein 17, V,1, 177
Fieser 02,389; 03,272; 04,468; 12,455
Merck Index 15,9086
Solubility Information Solubility in water: 130g/L (20°C). Other solubilities: soluble in ethanol, acetone, chloroform, benzene
Formula Weight 118.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Butadiene sulfone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0437269 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number107610025,107611000,107615000
Lot NumberA0446278 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number107610025,107611000,107615000
Lot NumberA0449238 Certificate TypeCertificate of Analysis Date16/01/2025 Catalog Number107610025,107611000,107615000
Lot NumberA0465870 Certificate TypeCertificate of Analysis Date03/09/2024 Catalog Number107610025,107611000,107615000
Lot NumberA0441365 Certificate TypeCertificate of Analysis Date02/05/2024 Catalog Number107610025,107611000,107615000
compliance-icons
Product Identifier
  • Butadiene sulfone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Butadiene sulfone, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.