missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Potent, selective non-peptide CCR1 antagonist (Ki = 1 nM for human CCR1). Exhibits 250-fold selectivity for CCR1 over CCR2, CCR5 and CXCR4. Inhibits MIP-α/CCL3-induced intracellular Ca2+ mobilization. Orally active; effectively reduces disease severity in a rat model of multiple sclerosis. Decreases renal fibrosis in a mouse model of obstructive nephropathy.

Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 434.89 |
Percent Purity | >98% |
Chemical Name or Material | BX 471 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BX 471, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction