missing translation for 'onlineSavingsMsg'
Learn More
Phenylurea, 97%, Thermo Scientific Chemicals Product Code.: 10205090

Phenylurea, 97%, Thermo Scientific Chemicals

Product Code. 10205090
100 g, Plastic bottle
Click to view available options
Quantity:
5 g
100 g
250 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10205090

Brand: Thermo Scientific Acros 131021000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-10-8
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00007944
InChI Key LUBJCRLGQSPQNN-UHFFFAOYSA-N
Synonym 1-phenylurea, n-phenylurea, urea, phenyl, phenylcarbamide, monophenylurea, stabilizer vh, stabilisator vh, phenyl-urea, urea, n-phenyl, phenyl ureaShow MoreShow Less
PubChem CID 6145
IUPAC Name phenylurea
SMILES C1=CC=C(C=C1)NC(=O)N

Specifications

Melting Point 143°C to 149°C
Color White to Yellow
Boiling Point 238°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula C6H5NHCONH2
Quantity 100 g
Beilstein 12, 346
Merck Index 15, 7429
Solubility Information Solubility in water: 1g/100mL. Other solubilities: soluble in hot alcohol,ether,ethylacetate,and glacial acetic acid
Formula Weight 136.15
Percent Purity 97%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Phenylurea, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0436140 Certificate TypeCertificate of Analysis Date04/03/2024 Catalog Number131022500,131021000
Lot NumberA0398098 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number131022500,131021000,131020050
Lot NumberA0406746 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number131022500,131021000,131020050
Lot NumberA0443117 Certificate TypeCertificate of Analysis Date14/06/2022 Catalog Number131022500,131021000,131020050
Lot NumberA0435097 Certificate TypeCertificate of Analysis Date05/10/2021 Catalog Number131022500,131021000,131020050
compliance-icons
Product Identifier
  • Phenylurea
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phenylurea, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.