Learn More
asecos™ Cabinet for Chemicals CS-CLASSIC
Safe and proper storage of non-flammable chemicals and toxic substances in working areas
812.00 € - 1695.00 €
Specifications
Certifications/Compliance | DIN EN 16121/16122 conformity |
---|---|
Depth (Metric) Exterior | 520 mm |
Depth (Metric) Interior | 493 mm |
Material | Steel |
Product Type | Cabinet for chemicals |
Alert:
For indoor use only
Product Code | Brand | Width (Metric) Exterior | Configuration | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) Exterior | Configuration | Color | Price | Quantity & Availability | |||
15915006
|
asecos™
3053004130534 |
810 mm | 2 x 60 kg load shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel galvanized) | Gray |
812.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15954996
|
asecos™
3053004030534 |
810 mm | 2 x 60 kg load shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel galvanized) | Anthracite Gray, Warning Yellow |
812.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15935006
|
asecos™
3053004130536 |
810 mm | 2 x 60 kg load shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Anthracite Gray, Light Gray |
829.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15974996
|
asecos™
3053004030536 |
810 mm | 2 x 60 kg load shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Anthracite Gray, Warning Yellow |
829.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15905116
|
asecos™
3053204130542 |
810 mm | 3 x 60 kg load shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel galvanized) | Anthracite Gray, Light Gray |
980.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15945106
|
asecos™
3053204030542 |
810 mm | 3 x 60 kg load shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel galvanized) | Anthracite Gray, Warning Yellow |
980.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15925116
|
asecos™
3053204130544 |
810 mm | 3 x 60 kg load shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Gray |
992.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15955106
|
asecos™
3053204030544 |
810 mm | 3 x 60 kg load shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Anthracite Gray, Warning Yellow |
992.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15985006
|
asecos™
3053004130541 |
810 mm | 3 x pull-out shelf (sheet steel powder-coated/PP) | Gray |
1060.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15516885
|
asecos™
3053004030541 |
810 mm | 3 x pull-out shelf (sheet steel powder-coated/PP) | Gray, Yellow |
1060.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15965116
|
asecos™
3053204130548 |
810 mm | 4 x pull-out shelf (sheet steel powder-coated/PP) | Anthracite Gray, Light Gray |
1335.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15526885
|
asecos™
3053204030548 |
810 mm | 4 x pull-out shelf (sheet steel powder-coated/PP) | Gray, Yellow |
1335.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15580051
|
asecos™
3053204130550 |
810 mm | 6 x pull-out shelf (sheet steel powder-coated/PP) | Gray |
1630.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15536885
|
asecos™
3053204030550 |
810 mm | 6 x pull-out shelf (sheet steel powder-coated/PP) | Gray, Yellow |
1630.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15593873
|
asecos™
3049204130498 |
1055 mm | 2 x 100 kg load shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel galvanized) | Gray |
846.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15914586
|
asecos™
3049204030498 |
1055 mm | 2 x 100 kg load shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel galvanized) | Gray, Yellow |
846.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15904596
|
asecos™
3049204130500 |
1055 mm | 2 x 100 kg load shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Gray |
863.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15934586
|
asecos™
3049204030500 |
1055 mm | 2 x 100 kg load shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Anthracite Gray, Warning Yellow |
863.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15974786
|
asecos™
3049504030510 |
1055 mm | 3 x 100 kg load shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Anthracite Gray, Warning Yellow |
1025.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15974596
|
asecos™
3049204130507 |
1055 mm | 3 x pull-out shelf (sheet steel powder-coated/PP) | Anthracite Gray, Light Gray |
1105.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15526875
|
asecos™
3049204030507 |
1055 mm | 3 x pull-out shelf (sheet steel powder-coated/PP) | Gray, Yellow |
1105.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15904806
|
asecos™
3049504130516 |
1055 mm | 4 x pull-out shelf (sheet steel powder-coated/PP) | Gray |
1385.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15914806
|
asecos™
3049504130518 |
1055 mm | 6 x pull-out shelf (sheet steel powder-coated/PP) | Gray |
1695.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15235403
|
asecos™
3049504030518 |
1055 mm | 6 x pull-out shelf (sheet steel powder-coated/PP) | Gray, Yellow |
1695.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated textured surface
- No unauthorised use: doors lockable with cylinder locking (integrated in the turning handle)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: natural ventilation due to ventilation openings at the cabinet bottom, ready for connection to a technical exhaust system
Specifications
DIN EN 16121/16122 conformity | |
493 mm | |
Cabinet for chemicals | |
Safe and proper storage of non-flammable chemicals and toxic substances in working areas | |
Powder-coated textured |
520 mm | |
Steel | |
Wing | |
2 |