Learn More
Calcium carbonate, 98%, Thermo Scientific Chemicals
Description
Manufacture of paint, rubber, plastics, etc. In food, cosmetics, and pharmaceuticals. As soil conditioner, neutralizer of surface waters, and as an industrial acid neutralizerIt is used medicinally as a calcium supplement or as an antacid. It is also used for industrial purposes. Calcium carbonate is the most widely used mineral in paper, plastics, paints and coatings industries. Hierarchical hollow calcium carbonate particles shows an useful application as anticancer drug carrier. It plays an essential role to catalyze alcoholysis of fats and oils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Manufacture of paint, rubber, plastics, etc
- In food, cosmetics, and pharmaceuticals
- As soil conditioner, neutralizer of surface waters, and as an industrial acid neutralizer

Specifications
Specifications
Melting Point | 800°C (decomposition) |
Color | White |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 98% |
Merck Index | 14,1657 |
Solubility Information | Practically insoluble in water; Soluble in diluted acids |
Formula Weight | 100.09 |
Percent Purity | 98% |
Odor | Odorless |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X07A004 | Certificate of Analysis | 13/11/2024 | 012365.A3,012365.36 |
W12K017 | Certificate of Analysis | 27/09/2024 | 012365.A3,012365.36 |
T15K045 | Certificate of Analysis | 29/07/2024 | 012365.A3,012365.36 |
M23K021 | Certificate of Analysis | 07/02/2024 | 012365.A3,012365.36 |
P10A035 | Certificate of Analysis | 21/09/2023 | 012365.A3,012365.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.