missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium hydroxide, 98%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 2850.0°C |
Melting Point | 580.0°C |
Color | White |
Physical Form | Amorphous Powder |
Quantity | 1 kg |
Assay Percent Range | 97.5% min. (Complexometry) |
Linear Formula | Ca(OH)2 |
Merck Index | 15, 1675 |
Solubility Information | Solubility in water: 1.65g/L (20°C). Other solubilities: soluble in acids (evolution of heat) and nh4cl sol, soluble in glycerol and sugar, insoluble in alcohol |
IUPAC Name | calcium dihydroxide |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0467923 | Certificate of Analysis | 31/01/2025 | 219180100,219180010,219181000,219180025,219185000 |
A0467176 | Certificate of Analysis | 28/10/2024 | 219180100,219180010,219181000,219180025,219185000 |
A0457434 | Certificate of Analysis | 03/11/2023 | 219180100,219180010,219181000,219180025,219185000 |
A0457218 | Certificate of Analysis | 25/10/2023 | 219180100,219180010,219181000,219180025,219185000 |
A0456734 | Certificate of Analysis | 06/10/2023 | 219180100,219180010,219181000,219180025,219185000 |
Safety and Handling


Product Identifier
- Calcium hydroxide
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Calcium hydroxide, 98%, extra pure, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction