Learn More
Biochrom Calibration Fluid for NanoVue Plus Spectrophotometer
Use Calibration Fluid for NanoVue Plus Spectrophotometer to drop-and-measure sample plates for accurate and reliable quantitation of nucleic acid and protein sample with this Uracil solution in Sodium Azide.
Brand: Biochrom 80-2140-09
This item is not returnable.
View return policy
Description
- Calibration Fluid for use with NanoVue Plus Spectrophotometer
Specifications
Pathlength calibration fluid | |
No Display | |
260 to 280 nm | |
NanoVue Plus Spectrophotometer |
Diode | |
Xenon Flash | |
UV-Visible Spectrophotometer for Quick and Accurate Quantitation of Nucleic Acids and Protein Samples |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling
Product Identifier
Signal Word
Hazard Category
- Pathlength Calibration Fluid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Product Content Correction
Product Title
Biochrom Calibration Fluid for NanoVue Plus Spectrophotometer > Calibration Fluid; For use with: NanoVue Plus Spectrophotometer
Spot an opportunity for improvement?Share a Content Correction