Learn More
Thermo Scientific™ 96 Well Plate, Poly-D-Lysine Coated Surface, Pack of 5
Color:Clear; Lid:With Lid
Brand: Thermo Scientific™ 152039
538.50 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available
- Aseptically Manufactured

Specifications
96F well plate, Poly-D-Lysine, clear, with lid | |
400 μL | |
Case of 20 | |
20 | |
Poly-D-Lysine | |
96 | |
8 x 12 | |
5/Pack, 20/Case | |
Flat |
Polystyrene | |
Microplate | |
5 | |
Clear | |
Aseptically Manufactured | |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
Nunc™ | |
Custom Barcoding Available |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.