Learn More
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Excellent for melting point analysis in the laboratory
Brand:  Hirschmann™ 9211512
 View more versions of this product
Product Code. 10613202
  • 107.00 € / Pack of 1000
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Capillary tubes
  • One end closed
  • Available in a wide range of diameters/lengths
Specifications
Specifications
Melting Point Determination Tube, One End Closed
1.15 mm
120 mm
4 x 250/Pk.