Learn More
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Excellent for melting point analysis in the laboratory
Brand:  Hirschmann™ 9211360
 View more versions of this product
Product Code. 10720233
  • 68.45 € / Pack of 1000
Explore more special offers
Description
Description
  • Capillary tubes
  • One end closed
  • Available in a wide range of diameters/lengths
Specifications
Specifications
Melting Point Determination Tube, One End Closed
0.95 mm
60 mm
10 x 100/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed > Length Tube: 60 mm, Diameter Inner: 0.95 mm, Quantity: 10 x 100/Pack