missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon tetrabromide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent

Specifications
Specifications
Melting Point | 88°C to 93°C |
Color | White |
Boiling Point | 190°C |
Assay Percent Range | 97.5% min. (On organic basis) (GC) |
Packaging | Glass Bottle |
Linear Formula | CBr4 |
Quantity | 5 g |
Beilstein | 01, 68 |
Fieser | 15,352; 16,366 |
Solubility Information | Solubility in water: insoluble |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0461277 | Certificate of Analysis | 25/03/2024 | 325150050,325155000,325151000 |
A0440219 | Certificate of Analysis | 22/09/2023 | 325150050,325155000,325151000 |
A0451863 | Certificate of Analysis | 02/05/2023 | 325150050,325155000,325151000 |
A0447102 | Certificate of Analysis | 31/10/2022 | 325150050,325155000,325151000 |
A0407003 | Certificate of Analysis | 28/03/2022 | 325150050,325155000,325151000 |
Safety and Handling


Product Identifier
- Carbon tetrabromide
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Carbon tetrabromide, 98%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction