missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ DMEM (Dulbecco's Modified Eagle's Medium)

Corning™ DMEM (Dulbecco's Modified Eagle's Medium) support cell culture growth and substitution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation. Products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis.
64.33 € - 707.23 €
Specifications
Concentration | 1 X |
---|---|
Form | Liquid |
For Use With (Application) | To assess growth promotion and cytotoxicity characteristics |
Endotoxin Level | <0.25EU/mL |
Content And Storage | 2 to 8°C |
Product Code | Brand | Quantity | With Additives | Without Additives | Product Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | With Additives | Without Additives | Product Type | Price | Quantity & Availability | ||
15303601
![]() |
Corning™
10-014-CM |
6 x 1 L | Glucose, Glutamine, Sodium Pyruvate | None | DMEM Media, 1 g/L Glucose |
224.50 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15323601
![]() |
Corning™
10-017-CM |
6 x 1 L | Glutamine, Glucose | No Sodium Pyruvate | DMEM Media, 4.5 g/L Glucose |
234.05 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15323531
![]() |
Corning™
10-013-CV |
6 x 500 mL | Glucose, Glutamine, Sodium Pyruvate | None | DMEM Media, 4.5 g/L Glucose |
64.33 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15373591
![]() |
Corning™
10-013-CM |
6 x 1 L | Glucose, Glutamine, Sodium Pyruvate | None | DMEM Media, 4.5 g/L Glucose |
248.19 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15353531
![]() |
Corning™
10-017-CV |
6 x 500 mL | Glucose, Glutamine | No Sodium Pyruvate | DMEM Media, 4.5 g/L Glucose |
70.65 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15353551
![]() |
Corning™
15-013-CV |
6 x 500 mL | Glucose, Sodium Pyruvate | No Glutamine | DMEM Media, 4.5 g/L Glucose |
78.54 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15383551
![]() |
Corning™
15-017-CV |
6 x 500 mL | Glucose | No Glutamine, No Sodium Pyruvate | DMEM Media, 4.5 g/L Glucose |
162.27 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15393551
![]() |
Corning™
15-018-CV |
6 x 500 mL | Glucose, Sodium Pyruvate, HEPES | No Glutamine | DMEM Media, 4.5 g/L Glucose |
194.78 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15383561
![]() |
Corning™
17-205-CV |
6 x 500 mL | Glucose, Sodium Pyruvate | No Glutamine, No Phenol Red | DMEM Media, 4.5 g/L Glucose |
173.63 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15393611
![]() |
Corning™
15-013-CM |
6 x 1 L | Glucose, Sodium Pyruvate | No Glutamine | DMEM Media, 4.5 g/L Glucose |
220.04 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15393591
|
Corning™
10-013-LX |
1 x 10 L | Glucose, Glutamine, Sodium Pyruvate | None | DMEM Media, 4.5 g/L Glucose |
358.65 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15313621
![]() |
Corning™
15-013-LX |
1 x 10 L | Glucose, Sodium Pyruvate | No Glutamine | DMEM Media, 4.5 g/L Glucose |
482.31 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15303621
![]() |
Corning™
15-013-LB |
1 x 20 L | Glucose, Sodium Pyruvate | No Glutamine | DMEM Media, 4.5 g/L Glucose |
707.23 €
20 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15333621
![]() |
Corning™
15-018-CM |
6 x 1 L | Glucose, Sodium Pyruvate, HEPES | No Glutamine | DMEM Media, 4.5 g/L Glucose |
328.09 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15323621
![]() |
Corning™
15-017-CM |
6 x 1 L | Glucose | No Glutamine, No Sodium Pyruvate | DMEM Media, 4.5 g/L Glucose |
231.94 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
- Original formulation was used to culture embryonic mouse cells
- Has been modified to support variety of normal and transformed cells, such as primary cultures of mouse and chicken cells
- Formulations can differ in amount or absence of components such as glucose, sodium pyruvate, sodium bicarbonate and L-glutamine
- Tested to determine pH and osmolality
- Low levels of endotoxin, and absence of contamination
- Tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) Chromogenic Assay
- Osmolality: 335±mOsm/kg H2O
- Mycoplasma: Tested Negative
- Sterility: Pass
Specifications
1 X | |
To assess growth promotion and cytotoxicity characteristics | |
2 to 8°C |
Liquid | |
<0.25EU/mL |
Safety and Handling
missing translation for 'shelfLife' : 18 Months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ DMEM (Dulbecco's Modified Eagle's Medium)