Learn More
Sartorius Cellulose Extraction Thimbles
Extraction thimbles been designed to be compatible with the Soxhlet apparatus, making them ideal for a variety of applications. Designed for versatility, the thimbles can be used with any extractor, including models like the Tectator, to facilitate efficient and high-yield extraction procedures.
118.00 € - 219.00 €
Product Code | Brand | Diameter (Metric) Inside | Length (Metric) | Wall Thickness | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Length (Metric) | Wall Thickness | Price | Quantity & Availability | ||||
30052893
|
Sartorius
FT-1201-019090 |
19 mm | 90 mm | 1.3 mm |
118.00 €
Pack of 25 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||
30052914
|
Sartorius
FT-1201-022080 |
22 mm | 80 mm | 1.3 mm |
118.00 €
Pack of 25 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||
30052895
|
Sartorius
FT-1201-022100 |
22 mm | 100 mm | 1.3 mm |
127.00 €
Pack of 25 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
30052920
|
Sartorius
FT-1201-025060 |
25 mm | 60 mm | 1.3 mm |
127.00 €
Pack of 25 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
30052910
|
Sartorius
FT-1201-025070 |
25 mm | 70 mm | 1.3 mm |
127.00 €
Pack of 25 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
30052897
|
Sartorius
FT-1201-025080 |
25 mm | 80 mm | 1.3 mm |
127.00 €
Pack of 25 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||
30052924
|
Sartorius
FT-1201-025100 |
25 mm | 100 mm | 1.3 mm |
127.00 €
Pack of 25 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||
30052921
![]() |
Sartorius
FT-1201-026060 |
26 mm | 60 mm | 1.5 mm |
127.00 €
Pack of 25 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||
30052899
|
Sartorius
FT-1201-028060 |
28 mm | 60 mm | 1.3 mm |
127.00 €
Pack of 25 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
30052913
|
Sartorius
FT-1201-028080 |
28 mm | 80 mm | 1.3 mm |
127.00 €
Pack of 25 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||
30052894
|
Sartorius
FT-1201-028100 |
28 mm | 100 mm | 1.3 mm |
127.00 €
Pack of 25 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||
30052922
|
Sartorius
FT-1201-030080 |
30 mm | 80 mm | 1.3 mm |
135.00 €
Pack of 25 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||
30052904
|
Sartorius
FT-1201-030100 |
30 mm | 100 mm | 1.3 mm |
135.00 €
Pack of 25 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||
30052919
|
Sartorius
FT-1201-033060 |
33 mm | 60 mm | 1.3 mm |
135.00 €
Pack of 25 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
30052912
![]() |
Sartorius
FT-1201-033080 |
33 mm | 80 mm | 1.3 mm |
135.00 €
Pack of 25 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||
30052909
![]() |
Sartorius
FT-1201-033090 |
33 mm | 90 mm | 1.3 mm |
135.00 €
Pack of 25 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||
30052892
![]() |
Sartorius
FT-1201-033094 |
33 mm | 94 mm | 1.3 mm |
135.00 €
Pack of 25 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||
30052915
![]() |
Sartorius
FT-1201-033100 |
33 mm | 100 mm | 1.3 mm |
141.00 €
Pack of 25 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||
30052917
![]() |
Sartorius
FT-1201-033118 |
33 mm | 118 mm | 1.3 mm |
141.00 €
Pack of 25 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||
30052896
![]() |
Sartorius
FT-1201-033130 |
33 mm | 130 mm | 1.5 mm |
141.00 €
Pack of 25 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||
30052898
![]() |
Sartorius
FT-1201-033205 |
33 mm | 205 mm | 1.3 mm |
208.00 €
Pack of 25 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||
30052907
![]() |
Sartorius
FT-1201-035150 |
35 mm | 150 mm | 1.7 mm |
208.00 €
Pack of 25 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||
30052902
![]() |
Sartorius
FT-1201-040100 |
40 mm | 100 mm | 1.7 mm |
219.00 €
Pack of 25 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
30052908
![]() |
Sartorius
FT-1201-040123 |
40 mm | 123 mm | 1.7 mm |
219.00 €
Pack of 25 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
30052900
![]() |
Sartorius
FT-1201-040150 |
40 mm | 150 mm | 1.8 mm |
219.00 €
Pack of 25 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||
30052923
![]() |
Sartorius
FT-1201-043123 |
43 mm | 123 mm | 1.5 mm |
219.00 €
Pack of 25 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||
Beschreibung
- Composed of high-purity cellulose for uncontaminated results
- Engineered for superior mechanical stability to withstand rigorous use
- Designed for a precise fit across a diverse range of extractors
- Exhibits outstanding chemical resistance, ensuring reliability in various extraction processes
Spezifikation
Cellulose | |
C300 | |
Extraction Thimbles |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Sartorius Cellulose Extraction Thimbles