Learn More
Macherey-Nagel™ Chromafix™ SB Cartridge
210.00 € - 316.00 €
Specifications
Base Material Silica
For Use With (Application) Organic Acids, Caffeine, Saccharin
Particle Size 45 μm
Sorbent SB
Type Chromafix SB Cartridge
View More Specs
Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10236571
View Documents
Macherey-Nagel™
731836
Adsorbent Weight: 920 mg
279.00 €
Pack of 50
In Stock
to see stock.
Add to basket
10458981
View Documents
Macherey-Nagel™
731834
Adsorbent Weight: 230 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10053051
View Documents
Macherey-Nagel™
731835
Adsorbent Weight: 460 mg
210.00 €
Pack of 50
In Stock
to see stock.
Add to basket
Specifications
Silica
45 μm
Chromafix SB Cartridge
50 Pack
Organic Acids, Caffeine, Saccharin
SB
60 Å
500 m2/g
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Macherey-Nagel™ Chromafix™ SB Cartridge