Learn More
Macherey-Nagel™ Chromafix™ SB Cartridge
Brand:  Macherey-Nagel™ 731835
 View more versions of this product
Product Code. 10053051
  • 210.00 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
Silica
Organic Acids, Caffeine, Saccharin
SB
60 Å
500 m2/g
Adsorbent Weight: 460 mg
45 μm
Chromafix SB Cartridge
50 Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Chromafix™ SB Cartridge > Adsorbent Weight: 460mg