Learn More
Macherey-Nagel™ Chromafix™ SB Cartridge
Brand: Macherey-Nagel™ 731836
Specifications
Silica | |
Organic Acids, Caffeine, Saccharin | |
SB | |
60 Å | |
500 m2/g |
Adsorbent Weight: 920 mg | |
45 μm | |
Chromafix SB Cartridge | |
50 Pack |
Product Suggestions
Customers who viewed this item also viewed