Learn More
Proteintech ChromoTek Nano-CaptureLigand™ Human IgE, Recombinant VHH, Biotinylated
Nano-CaptureLigand for human IgE: single domain antibody/VHH [CTK0111], biotinylated.
Brand:  Proteintech shuEB-1-10
 View more versions of this product
Product Code. 17333463
  • 113.00 € / 10µL
Explore more special offers
Description
Description
  • The ChromoTek Nano-CaptureLigand™ human IgE, VHH, biotinylated is used for the site-directed and specific immobilization of human IgE in biosensor and ELISA assays
  • It captures non-biotinylated human IgE antibodies to streptavidin/avidin
  • Nano-CaptureLigand human IgE, VHH, biotinylated comprises a monoclonal biotinylated VHH/ Nanobody
Specifications
Specifications
10 μL
Biotin
Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F
Fc-fragment of Human IgG1, IgG2, IgG3, and IgG4, Fc-fragment of Rabbit IgG
Monoclonal
BLI, SPR, ELISA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Proteintech ChromoTek Nano-CaptureLigand™ Human IgE, Recombinant VHH, Biotinylated > 10 μL