Learn More
Hirschmann™ Class A Measuring Cylinders, Schellbach Graduation
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class A Measuring Cylinders, Schellbach Graduation are manufactured from durable DURAN™ glass tubing.
32.00 € - 163.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10238283
|
Hirschmann™
2220160 |
10 mL |
32.00 €
Pack of 2 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
13157939
|
Hirschmann™
2220154 |
5 mL |
33.15 €
Pack of 2 |
Estimated Shipment: 07-01-2025
Log in to see stock. |
|||||
11857523
|
Hirschmann™
2220170 |
25 mL |
32.70 €
Pack of 2 |
Estimated Shipment: 10-12-2024
Log in to see stock. |
|||||
12678665
|
Hirschmann™
2220186 |
250 mL |
70.65 €
Pack of 2 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
11886983
|
Hirschmann™
2220175 |
50 mL |
34.85 €
Pack of 2 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
12668665
|
Hirschmann™
2220180 |
100 mL |
43.30 €
Pack of 2 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
11806993
|
Hirschmann™
2220190 |
500 mL |
119.00 €
Pack of 2 |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
10687321
|
Hirschmann™
2220193 |
1000 mL |
78.75 €
Each |
Estimated Shipment: 06-12-2024
Log in to see stock. |
|||||
10325611
|
Hirschmann™
2220194 |
2000 mL |
163.00 €
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
Description
- With spout, main point ring graduation
- Blue, Schellbach stain graduation
- Conformity certified, with dated batch certificate
- Acc. to DIN EN ISO 4788
- Corrosion-free and resistant to thermal shock
Specifications
DIN EN ISO 4788 | |
A |