Learn More
Sampling Systems™ Cleaning Coupons for Metal
Test and validate your cleaning regime with this range of cleaning validation coupons for various metals.
Brand:  Sampling Systems™ 8644S4-50B
 View more versions of this product
Product Code. 15899551
  • 158.00 € / Each
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Tests available for aluminum, 304 stainless steel and 316 stainless steel
  • All metals also available for either bright polish or satin finish
  • Two sizes available for each configuration, either 50 x 50mm or 100 x 100mm
Specifications
Specifications
Cleaning Coupon
Bright Polish
Stainless Steel (304)
50 x 50 mm
Testing and validating cleaning regime
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sampling Systems™ Cleaning Coupons for Metal > Material: 304 Stainless steel bright polish; Size: 50 x 50mm