Learn More
Sampling Systems™ Cleaning Coupons for Metal
Test and validate your cleaning regime with this range of cleaning validation coupons for various metals.
79.45 € - 196.00 €
Product Code | Brand | Material | Dimensions (L x W) | Finish | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Dimensions (L x W) | Finish | Price | Quantity & Availability | ||||
15889541
|
Sampling Systems™
8644AL-50 |
Aluminum | 50 x 50 mm | Bright Polish |
79.45 €
Each |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||
15899551
|
Sampling Systems™
8644S4-50B |
Stainless Steel (304) | 50 x 50 mm | Bright Polish |
158.00 €
Each |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||
15809561
|
Sampling Systems™
8644S4-50S |
Stainless Steel (304) | 50 x 50 mm | Satin finish |
90.10 €
Each |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||
15839561
|
Sampling Systems™
8644S6-50B |
Stainless Steel (316) | 50 x 50 mm | Bright Polish |
155.00 €
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
||||
15849561
|
Sampling Systems™
8644S6-50S |
Stainless Steel (316) | 50 x 50 mm | Satin finish |
128.00 €
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||
15879541
|
Sampling Systems™
8644AL-100 |
Aluminum | 100 x 100 mm | Bright Polish |
152.00 €
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||
15879551
|
Sampling Systems™
8644S4-100B |
Stainless Steel (304) | 100 x 100 mm | Bright Polish |
171.00 €
Each |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||
15889551
|
Sampling Systems™
8644S4-100S |
Stainless Steel (304) | 100 x 100 mm | Satin finish |
132.00 €
Each |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||
15819561
|
Sampling Systems™
8644S6-100B |
Stainless Steel (316) | 100 x 100 mm | Bright Polish |
196.00 €
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||
15829561
|
Sampling Systems™
8644S6-100S |
Stainless Steel (316) | 100 x 100 mm | Satin finish |
181.00 €
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||
Description
- Tests available for aluminum, 304 stainless steel and 316 stainless steel
- All metals also available for either bright polish or satin finish
- Two sizes available for each configuration, either 50 x 50mm or 100 x 100mm
Specifications
Cleaning Coupon |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Sampling Systems™ Cleaning Coupons for Metal