Learn More
Sampling Systems™ Cleaning Coupons for Metal
Test and validate your cleaning regime with this range of cleaning validation coupons for various metals.
Brand:  Sampling Systems™ 8644S4-100B
 View more versions of this product
Product Code. 15879551
  • 171.00 € / Each
Estimated Shipment: 08-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Tests available for aluminum, 304 stainless steel and 316 stainless steel
  • All metals also available for either bright polish or satin finish
  • Two sizes available for each configuration, either 50 x 50mm or 100 x 100mm
Specifications
Specifications
Cleaning Coupon
Bright Polish
Stainless Steel (304)
100 x 100 mm
Testing and validating cleaning regime
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sampling Systems™ Cleaning Coupons for Metal > Material: 304 Stainless steel bright polish; Size: 100 x 100mm