Learn More
Micronova™ Cleanroom Pens
BIOPHARMA
These low-sodium, indelible ink ballpoint pens feature a unique polypropylene barrel that limits particle generation and holds up to IPA, bleach and phenol wipe downs. Available irradiated, capped or click versions, and with black, blue or red ink.
Brand:  Micronova™ PEN-01
46.68 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.
 View more versions of this product
Product Code. 11992475
  • 52.20 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Low-sodium ink
The Micronova Cleanroom Ballpoint Pens, with low sodium ink and low particulate barrels, are designed for documentation and labeling in cleanrooms and aseptic environments. Gamma irradiated pens are suitable for Aseptic ISO 5 and other clean production areas.
  • Ball point cleanroom pens with indelible, low sodium ink
  • Designed for documentation and labeling in cleanrooms
  • White, high-density polypropylene barrel
  • Holds up to IPA, bleach and phenol wipe downs
  • Capped or Retractable options available
  • Black, blue or red ink
  • Available gamma irradiated for aseptic environments
Specifications
Specifications
HDPE
Capped
Not Autoclavable
Cleanroom and Aseptic Environment Documentation and Labeling
Non-sterile
Black
5.75 in.
Reusable
Cleanroom Pen
0.7 mm Point Tip
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Micronova™ Cleanroom Pens > Cleanroom Pen, ball-point pen low sodium Ink, black