Learn More
Corning™ Clear Polystyrene 96-Well EIA/RIA Microplates

Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.
Brand: Corning™ 9018
246.32 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
- Cell culture microplates are sterile and nonpyrogenic.
- Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.

Specifications
High Binding | |
Without Lid | |
25/Pk. | |
Clear | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
100/Cs. | |
100/Cs. |
Standard | |
8 x 12 | |
High Bind | |
Polystyrene | |
75 to 200 μL | |
High Binding | |
Capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions. | |
25/Pk. |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.