missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Yeast Extract (Granulated)
Dried yeast autolysate with a high content of amino nitrogen and water soluble B complex vitamins
Brand: Fisher BioReagents™ BP9727-2
This item is not returnable.
View return policy
Description
Dried yeast autolysate with a high content of amino nitrogen and water soluble B complex vitamins
- Water-soluble portion of autolyzed yeast containing vitamin B complex
- Used in preparing microbiological culture media
- CAS: 8013-01-2

Specifications
Dehydrated | |
Bacterial Culture | |
Vitamin B complex | |
2000 g |
Yeast Extract | |
Microbiological Culture Media | |
Poly Bottle |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Safety and Handling
Product Identifier
- Yeast Extract
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisher BioReagents™ Yeast Extract (Granulated) > 2kg
Spot an opportunity for improvement?Share a Content Correction