missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CO2 Independent Medium
GREENER_CHOICE

A non-HEPES proprietary medium used for supporting cell growth for a variety of suspension and adherent mammalian cells.

Brand:  Gibco™ 18045088

97.34 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 15420604

  • 106.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • CO2 Independent Medium is a non-HEPES proprietary medium used for supporting cell growth for a variety of suspension and adherent mammalian cells such as epithelial, fibroblast, and lymphoid cell lines without a CO2 incubator
  • CO2 Independent Medium is ideal for transporting cells or tissue, and for the handling of mouse embryos under atmospheric conditions
  • Gibco™ CO2 Independent Medium contains a unique buffering system composed of mono and dibasic sodium phosphate and β-glycerophosphate
  • A small amount of sodium bicarbonate has been included in the formulation to meet essential bicarbonate dependent functions
  • No synthetic buffers are utilized, thus eliminating any cytotoxic effects associated with such buffering systems
  • Additionally, Gibco™ CO2 Independent Medium has been formulated with components that enhance cellular production and utilization of CO2
  • Store at 2°C to 8°C, protected from light

Modified as follows

  • With: phenol red, sodium pyruvate
  • Without: L-glutamine, HEPES

Using CO2 Independent Medium

  • CO2 Independent Medium contains no lipids or growth factors therefore it requires supplementation, commonly with 10% Fetal Bovine Serum (FBS)
  • The CO2 Independent Medium does not require a CO2 environment to maintain physiological pH
  • For maximum growth performance some cell lines may require either direct or sequential adaptation to CO2 Independent Medium
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Epithelial, fibroblast, and lymphoid
Liquid
Sterile-filtered
No Glutamine, No HEPES
Room Temperature
Mouse Embryos
1 X
CO2 Independent Medium
Phenol Red, Sodium Pyruvate
500 mL
Sustainable packaging
Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2753834 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number18045088
Lot Number2415508 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number18045088
Lot Number2473962 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number18045088
Lot Number2481591 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number18045088
Lot Number2517980 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number18045088
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ CO2 Independent Medium > 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.