Learn More
Nabertherm™ Compact Tube Furnace with B510 Controller
These compact tube furnaces with integrated control systems can be used universally for many processes. Equipped with a standard working tube of C 530 ceramic and two fiber plugs, these tube furnaces have an unbeatable price/performance ratio.
5960.00 € - 12170.00 €
Specifications
Controller Type B510
Type Compact Tube Furnace
View More Specs
Products 6
Product Code Brand Diameter (Metric) Process Tube Heated Zone Temperature (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Process Tube Heated Zone Temperature (Metric) Price Quantity & Availability  
17318089
View Documents
Nabertherm™
R-322L2AN
120 mm 500 mm 1200°C
5960.00 €
Each
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
17328089
View Documents
Nabertherm™
R-322L3AN
120 mm 500 mm 1300°C
6765.00 €
Each
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
17388089
View Documents
Nabertherm™
R-472L3AN
100 mm 750 mm 1300°C
10230.00 €
Each
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
17378089
View Documents
Nabertherm™
R-472L2AN
170 mm 750 mm 1200°C
9045.00 €
Each
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
17338099
View Documents
Nabertherm™
R-572L2AN
170 mm 1000 mm 1200°C
10675.00 €
Each
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
17348099
View Documents
Nabertherm™
R-572L3AN
170 mm 1000 mm 1300°C
12170.00 €
Each
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
Description
Description
  • Excellent price - performance ratio
  • Can also be equipped with gas supply system 1 (no vacuum operation) and gas supply system 2 (for vacuum operation)
  • Temperature, max. 1200°C or 1300°C
  • Single-zoned design as standard with controller B 510
  • Outer tube diameter of 50 to 170 mm, heated length from 250 to 1000 mm
  • Dual shell housing made of sheets of textured stainless steel
  • Insulation made of non-classified fibre material
Specifications
Specifications
B510
Compact Tube Furnace
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Nabertherm™ Compact Tube Furnace with B510 Controller