Learn More
Invitrogen™ Concanavalin A (Con A) Conjugates
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates.
Brand: Invitrogen™ C11253
241.50 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Description
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates. Concanavalin A (Con A) has been used in a variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC) and flow cytometry (FC). In many cell types, concanavalin A (Con A) is a good marker of ER and Golgi.
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Flour and Alexa Flour Plus conjugates. Concanavalin A selectively binds to alpha‐mannopyranosyl and alpha‐glucopyranosyl residues. In neutral and alkaline solutions, concanavalin A exists as a tetramer with a molecular weight of approximately 104,000 Daltons. In acidic solutions (pH below 5.0), concanavalin A exists as a dimer. Conjugated concanavalin A has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).
Specifications
Immunocytochemistry, Immunohistochemistry, Immunofluorescence | |
Canavlia ensiformis | |
α-man, α-glc | |
Native | |
Store in freezer (-5 to -30°C) and protect from light. | |
Alexa Fluor Dyes | |
Natural | |
Lyophilized | |
Imaging | |
Alexa Fluor™ | |
Lectin |
5 mg | |
Alexa Fluor 594 | |
C. ensiformis | |
None | |
590/617 nm | |
Room Temperature | |
Lectins | |
Concanavalin A (Con A) | |
Red | |
Imaging Quality |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.